Assay Details
Gene Symbol: Rap1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005765.1 | NP_001005765.1 | 2 - 3 | 171 | 145 | |
GenBank mRNA | BC083813.1 | - | 2 - 3 | 171 | 7110254 | 145 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC083813.1,NM_001005765.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
