Assay Details
| Gene Symbol: Ciart | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134593.1 | NP_001128065.1 | 4 - 5 | 655 | 80 | |
| XM_008761317.1 | XP_008759539.1 | 9 - 10 | 3126 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ciart
Genomic Map