Assay Details
| Gene Symbol: Lrig2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107710.1 | NP_001101180.1 | 17 - 18 | 3166 | 87 | |
| XM_006233083.2 | XP_006233145.1 | 17 - 18 | 3540 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P059237 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P059242 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768512 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868695 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 484656 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106567 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 777687 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869681 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 902917 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105146 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863092 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323023 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257178 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 36719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162300 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 870119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686451 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 108057 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563618 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433537 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 285031 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93316 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 527279 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 820874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660038 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617603 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561886 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529428 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643813 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 830009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816962 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130087 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 16391 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174444 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94459 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34934 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126891 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P059237 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P059242 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P059237 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P059242 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P059242 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P059237 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P356445 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 40140 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29067 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10825651 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825637 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825647 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825642 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825646 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825653 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825643 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825649 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825638 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825640 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825639 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825650 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825652 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825644 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825654 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825645 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825648 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10825641 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10825636 | - | - |
| Affymetrix Rat Clariom S Assay | PSR0200030181.rn.1 | TC0200004372.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0200030180.rn.1 | TC0200004372.rn.1 | RTA TC |
| Affymetrix Rat Gene 1.0 ST Array | 10825636 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21752565 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1351632 | - | - |

Lrig2
Genomic Map