Assay Details
Gene Symbol: Phgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031620.1 | NP_113808.1 | 11 - 12 | 1323 | 103 | |
XM_006233010.1 | XP_006233072.1 | 10 - 11 | 1346 | 103 | ||
GenBank mRNA | BC086327.1 | - | 10 - 11 | 1351 | 7115765 | 103 |
X97772.1 | - | 10 - 11 | 1282 | 103 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
