Assay Details
| Gene Symbol: Igsf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106455.1 | NP_001099925.1 | 6 - 7 | 2064 | 60 | |
| XM_006233041.2 | XP_006233103.1 | 8 - 9 | 2867 | 60 | ||
| XM_006233042.2 | XP_006233104.1 | 8 - 9 | 2864 | 60 | ||
| XM_006233043.2 | XP_006233105.1 | 7 - 8 | 2807 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igsf3
Genomic Map