Assay Details
| Gene Symbol: Mov10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107711.1 | NP_001101181.1 | 4 - 5 | 1000 | 70 | |
| XM_006233085.1 | XP_006233147.1 | 4 - 5 | 888 | 70 | ||
| XM_008761395.1 | XP_008759617.1 | 4 - 5 | 1748 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mov10
Genomic Map