Assay Details
| Gene Symbol: Mrps21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001287116.1 | NP_001274045.1 | 1 - 2 | 51 | 103 | |
| GenBank mRNA | BC166891.1 | - | 1 - 2 | 51 | 6918483 | 103 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC166891.1,NM_001287116.1 transcripts.
This assay is in the 5'UTR region of the BC166891.1,NM_001287116.1 transcripts.
Set Membership: |
|

Mrps21
Genomic Map