Assay Details
| Gene Symbol: Car14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109655.2 | NP_001103125.1 | 2 - 3 | 78 | 91 | |
| XM_006232988.2 | XP_006233050.1 | 3 - 4 | 623 | 152 | ||
| XM_006232989.2 | XP_006233051.1 | 3 - 4 | 601 | 91 | ||
| GenBank mRNA | EF187253.1 | - | 2 - 3 | 78 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006232988.2 transcripts.
Set Membership: |
|

Car14
Genomic Map