Assay Details
| Gene Symbol: Car14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109655.2 | NP_001103125.1 | 7 - 8 | 722 | 84 | |
| XM_006232988.2 | XP_006233050.1 | 8 - 9 | 1267 | 84 | ||
| XM_006232989.2 | XP_006233051.1 | 8 - 9 | 1245 | 84 | ||
| XM_006232990.2 | XP_006233052.1 | 8 - 9 | 1227 | 84 | ||
| GenBank mRNA | EF187253.1 | - | 7 - 8 | 722 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Car14
Genomic Map