Assay Details
| Gene Symbol: Mipep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031052.1 | NP_112314.1 | 1 - 2 | 270 | 58 | |
| XM_008770779.1 | XP_008769001.1 | 1 - 2 | 420 | 58 | ||
| XM_008770780.1 | XP_008769002.1 | 1 - 2 | 420 | 58 | ||
| GenBank mRNA | M96633.1 | - | 1 - 2 | 270 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mipep
Genomic Map