Assay Details
Gene Symbol: Cdca2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107273.1 | NP_001100743.1 | 14 - 15 | 1937 | 106 | |
XM_006252117.2 | XP_006252179.1 | 14 - 15 | 2014 | 106 | ||
XM_006252118.2 | XP_006252180.1 | 14 - 15 | 2122 | 106 | ||
XM_006252120.2 | XP_006252182.1 | 14 - 15 | 2011 | 106 | ||
XM_006252121.2 | XP_006252183.1 | 11 - 12 | 2068 | 106 | ||
XM_008770757.1 | XP_008768979.1 | 11 - 12 | 1394 | 106 | ||
GenBank mRNA | FQ211026.1 | - | 14 - 15 | 1921 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P478114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 572403 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968825 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 191824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 560269 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454748 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 160801 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796164 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740039 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574887 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 664322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1173 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 467193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 781577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874895 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59187 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119568 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57520 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P478114 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P478114 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P478114 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P478114 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 17958 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 94049 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10784731 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784740 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784742 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784736 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784733 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784735 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784737 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784739 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784744 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784738 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784741 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784730 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784732 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784743 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784734 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784746 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784745 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10784729 | - | - |
Rat Genome 230 2.0 Array | 1381833_at | - | - |
Rat Genome 230 2.0 Array | 1383958_PM_at | - | - |
Rat Genome 230 2.0 Array | 1383958_at | - | - |
Rat Genome 230 2.0 Array | 1381833_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1383958_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1381833_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1383958_at | - | - |
Affymetrix HT Rat Focus Array | 1381833_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1500012635.rn.1 | TC1500001875.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500012638.rn.1 | TC1500001875.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500012636.rn.1 | TC1500001875.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500012637.rn.1 | TC1500001875.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500012633.rn.1 | TC1500001875.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1500012634.rn.1 | TC1500001875.rn.1 | RTA TC |
Rat Expression Set 230 | 1383958_PM_at | - | - |
Rat Expression Set 230 | 1381833_at | - | - |
Rat Expression Set 230 | 1381833_PM_at | - | - |
Rat Expression Set 230 | 1383958_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10784729 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21842660 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371588 | - | - |
