Assay Details
| Gene Symbol: Gulo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022220.2 | NP_071556.2 | 7 - 8 | 746 | 74 | |
| XM_006252146.2 | XP_006252208.1 | 7 - 8 | 887 | 74 | ||
| GenBank mRNA | BC089803.1 | - | 7 - 8 | 746 | 7324344 | 74 |
| FQ219140.1 | - | 7 - 8 | 771 | 74 | ||
| FQ219632.1 | - | 7 - 8 | 854 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gulo
Genomic Map