Assay Details
| Gene Symbol: Gulo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022220.2 | NP_071556.2 | 9 - 10 | 978 | 79 | |
| GenBank mRNA | BC089803.1 | - | 9 - 10 | 978 | 7324344 | 79 |
| FQ219140.1 | - | 9 - 10 | 1003 | 79 | ||
| FQ219632.1 | - | 9 - 10 | 1086 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gulo
Genomic Map