Assay Details
Gene Symbol: Sh3rf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198764.1 | NP_942059.1 | 8 - 9 | 1802 | 78 | |
XM_008771225.1 | XP_008769447.1 | 7 - 8 | 1919 | 78 | ||
GenBank mRNA | AF515735.1 | - | 8 - 9 | 1802 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
