Assay Details
| Gene Symbol: Zfp386 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019620.2 | NP_062566.2 | 1 - 2 | 93 | 86 | |
| XM_006240685.2 | XP_006240747.1 | 1 - 2 | 145 | 86 | ||
| GenBank mRNA | BC078841.1 | - | 1 - 2 | 92 | 7115131 | 86 |
| U67082.1 | - | 1 - 2 | 82 | 86 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC078841.1, NM_019620.2, U67082.1, XM_006240685.2 transcripts.
Set Membership: |
|

Zfp386
Genomic Map