Assay Details
| Gene Symbol: Zfp952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002729770.4 | XP_002729816.1 | 3 - 4 | 2336 | 112 | |
| XM_008765188.1 | XP_008763410.1 | 4 - 5 | 478 | 112 | ||
More Information
Set Membership: |
|

Zfp952
Genomic Map