Assay Details
| Gene Symbol: Ncoa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276714.1 | NP_001263643.1 | 3 - 4 | 515 | 67 | |
| XM_006235296.2 | XP_006235358.1 | 5 - 6 | 852 | 67 | ||
| XM_006235297.1 | XP_006235359.1 | 5 - 6 | 749 | 67 | ||
| XM_008762295.1 | XP_008760517.1 | 6 - 7 | 921 | 67 | ||
| XM_008762296.1 | XP_008760518.1 | 5 - 6 | 749 | 67 | ||
| XM_008762297.1 | XP_008760519.1 | 5 - 6 | 749 | 67 | ||
| XM_008762298.1 | XP_008760520.1 | 5 - 6 | 749 | 67 | ||
| XM_008762299.1 | XP_008760521.1 | 5 - 6 | 749 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ncoa6
Genomic Map