Assay Details
Gene Symbol: Rem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025753.1 | NP_001020924.1 | 2 - 3 | 592 | 99 | |
XM_006235278.2 | XP_006235340.1 | 2 - 3 | 734 | 99 | ||
GenBank mRNA | BC098946.1 | - | 2 - 3 | 592 | 7460067 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
