Assay Details
Gene Symbol: Rem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025753.1 | NP_001020924.1 | 3 - 4 | 674 | 89 | |
XM_006235278.2 | XP_006235340.1 | 3 - 4 | 816 | 89 | ||
GenBank mRNA | BC098946.1 | - | 3 - 4 | 674 | 7460067 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
