Assay Details
| Gene Symbol: Cbfa2t2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168542.1 | NP_001162014.1 | 8 - 9 | 1348 | 62 | |
| XM_006235311.2 | XP_006235373.1 | 8 - 9 | 1332 | 62 | ||
| XM_006235312.2 | XP_006235374.1 | 9 - 10 | 1360 | 62 | ||
| XM_008762313.1 | XP_008760535.1 | 8 - 9 | 1436 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cbfa2t2
Genomic Map