Assay Details
| Gene Symbol: Csnk2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053824.2 | NP_446276.1 | 12 - 13 | 1156 | 77 | |
| XM_006235220.2 | XP_006235282.1 | 11 - 12 | 1216 | 77 | ||
| XM_006235221.2 | XP_006235283.1 | 11 - 12 | 1257 | 77 | ||
| GenBank mRNA | BC091130.1 | - | 12 - 13 | 1156 | 7367296 | 77 |
| L15618.1 | - | 1773 | 77 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
positive regulation of cell proliferation
Wnt signaling pathway
positive regulation of Wnt signaling pathway
positive regulation of cell growth
regulation of cell proliferation
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of protein catabolic process
protein autophosphorylation
rhythmic process
regulation of catalytic activity
protein serine/threonine kinase activity
ATP binding
beta-catenin binding
kinase activity
protein phosphatase regulator activity
protein N-terminus binding
regulation of cell growth
transcription, DNA-templated
regulation of transcription, DNA-templated
protein phosphorylation
positive regulation of cell proliferation
Wnt signaling pathway
positive regulation of Wnt signaling pathway
positive regulation of cell growth
regulation of cell proliferation
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
positive regulation of protein catabolic process
protein autophosphorylation
rhythmic process
regulation of catalytic activity
protein serine/threonine kinase activity
ATP binding
beta-catenin binding
kinase activity
protein phosphatase regulator activity
protein N-terminus binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P363204 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303492 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 569794 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59750 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 111913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162231 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798588 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 167337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941467 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428718 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236452 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 210397 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952743 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 1638 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308648 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652956 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459861 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58790 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714321 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 151623 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639132 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 851388 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 620529 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164822 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655624 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 322007 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818764 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58932 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12633 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 32816 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70517 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P363204 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P363204 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P363204 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P363204 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 700640577 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701245002 | - | - |
| Matsumoto (Agilent Rat cDNA Microarray G4105A) | 701643742 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 34954 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 90461 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 57019 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 100649 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701643742 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 701245002 | - | - |
| Agilent Rat cDNA Microarray (G4105A) | 700640577 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10840806 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840816 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840808 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840814 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840810 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840804 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840813 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840807 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840805 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840815 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840811 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840809 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840812 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10840803 | - | - |
| Rat Genome 230 2.0 Array | 1376616_at | - | - |
| Rat Genome 230 2.0 Array | 1387170_at | - | - |
| Rat Genome 230 2.0 Array | 1389974_at | - | - |
| Rat Genome 230 2.0 Array | 1392650_at | - | - |
| Rat Genome 230 2.0 Array | 1389974_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1387170_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1392650_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1375636_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1384339_PM_s_at | - | - |
| Rat Genome 230 2.0 Array | 1375636_at | - | - |
| Rat Genome 230 2.0 Array | 1384339_s_at | - | - |
| Rat Genome 230 2.0 Array | 1376616_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392650_at | - | - |
| Affymetrix HT Rat Focus Array | 1387170_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392650_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1387170_at | - | - |
| Affymetrix HT Rat Focus Array | 1384339_s_at | - | - |
| Affymetrix HT Rat Focus Array | 1384339_PM_s_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300014460.rn.1 | TC0300002028.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0300014459.rn.1 | TC0300002028.rn.1 | RTA TC |
| Rat Expression Set 230 | 1387170_PM_at | - | - |
| Rat Expression Set 230 | 1387170_at | - | - |
| Rat Expression Set 230 | 1384339_s_at | - | - |
| Rat Expression Set 230 | 1384339_PM_s_at | - | - |
| Rat Expression Set 230 | 1392650_at | - | - |
| Rat Expression Set 230 | 1392650_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840803 | - | - |
| Rat Genome U34 Set | L15618_at | - | - |
| Rat Genome U34 Set | rc_AI072089_at | - | - |
| Rat Genome U34 Set | rc_AI012245_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 22385428 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1363262 | - | - |
| RatRef-12 Expression BeadChip | ILMN_68679 | - | - |

Csnk2a1
Genomic Map