Assay Details
| Gene Symbol: Csnk2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053824.2 | NP_446276.1 | 7 - 8 | 607 | 111 | |
| XM_006235220.2 | XP_006235282.1 | 6 - 7 | 667 | 111 | ||
| XM_006235221.2 | XP_006235283.1 | 6 - 7 | 708 | 111 | ||
| GenBank mRNA | BC091130.1 | - | 7 - 8 | 607 | 7367296 | 111 |
| L15618.1 | - | 1224 | 111 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Csnk2a1
Genomic Map