Assay Details
| Gene Symbol: Hm13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107789.1 | NP_001101259.1 | 1 - 2 | 296 | 148 | |
| XM_006235268.1 | XP_006235330.1 | 1 - 2 | 307 | 148 | ||
| XM_006235269.1 | XP_006235331.1 | 1 - 2 | 307 | 148 | ||
| XM_008762330.1 | XP_008760552.1 | 1 - 2 | 307 | 148 | ||
More Information
Set Membership: |
|

Hm13
Genomic Map