Assay Details
Gene Symbol: Cst7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235213.2 | XP_006235275.1 | 2 - 3 | 1478 | 149 | |
XM_008762305.1 | XP_008760527.1 | 2 - 3 | 1478 | 149 | ||
XM_008762306.1 | XP_008760528.1 | 2 - 3 | 1475 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
