Assay Details
| Gene Symbol: Cnbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008762318.1 | XP_008760540.1 | 11 - 12 | 1561 | 72 | |
| XM_008762319.1 | XP_008760541.1 | 10 - 11 | 1504 | 72 | ||
| XM_008762320.1 | XP_008760542.1 | 9 - 10 | 1534 | 72 | ||
| XM_008762321.1 | XP_008760543.1 | 11 - 12 | 1561 | 72 | ||
| GenBank mRNA | AY539878.1 | - | 11 - 12 | 1789 | 72 | |
More Information
Set Membership: |
|

Cnbd2
Genomic Map