Assay Details
Gene Symbol: LOC100910990 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006235456.2 | XP_006235518.1 | 2 - 3 | 255 | 85 | |
XM_006235457.2 | XP_006235519.1 | 2 - 3 | 223 | 85 | ||
XM_008762413.1 | XP_008760635.1 | 1 - 2 | 117 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006235456.2, XM_006235457.2, XM_008762413.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
