Assay Details
| Gene Symbol: Tm9sf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025649.1 | NP_001020820.1 | 17 - 18 | 2000 | 79 | |
| XM_006235284.2 | XP_006235346.1 | 17 - 18 | 2018 | 79 | ||
| GenBank mRNA | BC099133.1 | - | 17 - 18 | 2000 | 7454474 | 79 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1032487 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544775 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57949 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286565 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842349 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783337 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491935 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53700 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310720 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717076 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889703 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 605149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 178835 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271408 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653127 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889658 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686855 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 896360 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652564 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547911 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 313434 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610069 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 717359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792411 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9426 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715631 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926838 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 443912 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 56533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91783 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 132684 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137532 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48555 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131933 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1032487 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1032487 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1032487 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P1032487 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 84282 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 68737 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 76396 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10840996 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841006 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841008 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840993 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841010 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840998 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840997 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841001 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841003 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840994 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841009 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841002 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840992 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840999 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841007 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841005 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840995 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840991 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10841000 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10840990 | - | - |
| Rat Genome 230 2.0 Array | 1367513_at | - | - |
| Rat Genome 230 2.0 Array | 1378075_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1367513_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1378052_at | - | - |
| Rat Genome 230 2.0 Array | 1394808_at | - | - |
| Rat Genome 230 2.0 Array | 1378052_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1378075_at | - | - |
| Rat Genome 230 2.0 Array | 1394808_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1394808_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1367513_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1394808_at | - | - |
| Affymetrix HT Rat Focus Array | 1378075_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1378075_at | - | - |
| Affymetrix HT Rat Focus Array | 1367513_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0300014808.rn.1 | TC0300002059.rn.1 | RTA TC |
| Rat Expression Set 230 | 1367513_PM_at | - | - |
| Rat Expression Set 230 | 1367513_at | - | - |
| Rat Expression Set 230 | 1394808_at | - | - |
| Rat Expression Set 230 | 1367513_at | - | - |
| Rat Expression Set 230 | 1378075_at | - | - |
| Rat Expression Set 230 | 1378075_PM_at | - | - |
| Rat Expression Set 230 | 1394808_PM_at | - | - |
| Rat Expression Set 230 | 1367513_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10840990 | - | - |
| Rat Genome U34 Set | rc_AI030680_at | - | - |
| Rat Genome U34 Set | rc_AA944528_g_at | - | - |
| Rat Genome U34 Set | rc_AA944528_at | - | - |
| Rat Genome U34 Set | rc_AI231294_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21844065 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374612 | - | - |

Tm9sf4
Genomic Map