Assay Details
| Gene Symbol: Ces2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100477.1 | NP_001093947.1 | 7 - 8 | 1054 | 101 | |
| GenBank mRNA | D50580.1 | - | 7 - 8 | 1046 | 101 | |
| FQ218265.1 | - | 7 - 8 | 1094 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ces2e
Genomic Map