Assay Details
| Gene Symbol: Ccl22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_057203.1 | NP_476551.1 | 1 - 2 | 95 | 69 | |
| GenBank mRNA | AF163477.1 | - | 1 - 2 | 40 | 69 | |
| BC088218.1 | - | 1 - 2 | 95 | 7384271 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccl22
Genomic Map