Assay Details
| Gene Symbol: Cdh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107407.1 | NP_001100877.1 | 11 - 12 | 1871 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel maturation
homophilic cell adhesion via plasma membrane adhesion molecules
negative regulation of cell proliferation
single organismal cell-cell adhesion
regulation of establishment of cell polarity
receptor binding
calcium ion binding
beta-catenin binding
protein phosphatase binding
ion channel binding
blood vessel maturation
homophilic cell adhesion via plasma membrane adhesion molecules
negative regulation of cell proliferation
single organismal cell-cell adhesion
regulation of establishment of cell polarity
receptor binding
calcium ion binding
beta-catenin binding
protein phosphatase binding
ion channel binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P145597 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710944 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656394 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 280892 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841710 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793049 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608630 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290486 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885594 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 676358 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254308 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586938 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69706 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711793 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795420 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432640 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 321006 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969197 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856191 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630685 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907682 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694471 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331693 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52249 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140149 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117244 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P145597 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P145597 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P145597 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P121658 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P19735 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 80669 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58466 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10809055 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809049 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809057 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809052 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809053 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809051 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809058 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809054 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809050 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809056 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809059 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10809048 | - | - |
| Rat Genome 230 2.0 Array | 1389255_PM_at | - | - |
| Rat Genome 230 2.0 Array | 1389255_at | - | - |
| Affymetrix HT Rat Focus Array | 1389255_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1389255_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR1900007435.rn.1 | TC1900000933.rn.1 | RTA TC |
| Rat Expression Set 230 | 1389255_PM_at | - | - |
| Rat Expression Set 230 | 1389255_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10809048 | - | - |
| Rat Genome U34 Set | rc_AA997619_at | - | - |
| Rat Genome U34 Set | rc_AA850824_at | - | - |
| Rat Genome U34 Set | rc_AA850824_g_at | - | - |
| Rat Genome U34 Set | rc_AI175977_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21684820 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359811 | - | - |

Cdh5
Genomic Map