Assay Details
| Gene Symbol: Setd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106167.1 | NP_001099637.1 | 3 - 4 | 695 | 95 | |
| XM_006255070.2 | XP_006255132.1 | 3 - 4 | 1573 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Setd6
Genomic Map