Assay Details
Gene Symbol: Setd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106167.1 | NP_001099637.1 | 5 - 6 | 1002 | 54 | |
XM_006255070.2 | XP_006255132.1 | 6 - 7 | 1952 | 54 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
