Assay Details
| Gene Symbol: Mt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138826.4 | NP_620181.1 | 1 - 2 | 98 | 89 | |
| GenBank mRNA | BC058442.1 | - | 1 - 2 | 103 | 6891030 | 89 |
| FQ209933.1 | - | 1 - 2 | 99 | 89 | ||
| FQ210006.1 | - | 1 - 2 | 99 | 89 | ||
| FQ210086.1 | - | 1 - 2 | 99 | 89 | ||
| FQ210225.1 | - | 1 - 2 | 99 | 89 | ||
| FQ210276.1 | - | 1 - 2 | 99 | 89 | ||
| FQ210404.1 | - | 1 - 2 | 89 | |||
| FQ210574.1 | - | 1 - 2 | 99 | 89 | ||
| FQ210578.1 | - | 1 - 2 | 99 | 89 | ||
| FQ210592.1 | - | 1 - 2 | 99 | 89 | ||
| FQ210741.1 | - | 1 - 2 | 99 | 89 | ||
| FQ210753.1 | - | 1 - 2 | 99 | 89 | ||
| FQ210805.1 | - | 1 - 2 | 99 | 89 | ||
| FQ210924.1 | - | 1 - 2 | 99 | 89 | ||
| FQ210951.1 | - | 1 - 2 | 99 | 89 | ||
| FQ211146.1 | - | 1 - 2 | 99 | 89 | ||
| FQ211312.1 | - | 1 - 2 | 99 | 89 | ||
| FQ211350.1 | - | 1 - 2 | 99 | 89 | ||
| FQ217792.1 | - | 1 - 2 | 99 | 89 | ||
| FQ218077.1 | - | 1 - 2 | 99 | 89 | ||
| FQ218157.1 | - | 1 - 2 | 99 | 89 | ||
| FQ218299.1 | - | 1 - 2 | 99 | 89 | ||
| FQ218315.1 | - | 1 - 2 | 99 | 89 | ||
| FQ220411.1 | - | 1 - 2 | 99 | 89 | ||
| FQ221163.1 | - | 1 - 2 | 99 | 89 | ||
| FQ221173.1 | - | 1 - 2 | 99 | 89 | ||
| FQ221343.1 | - | 1 - 2 | 99 | 89 | ||
| FQ221498.1 | - | 1 - 2 | 99 | 89 | ||
| FQ221507.1 | - | 1 - 2 | 99 | 89 | ||
| FQ221511.1 | - | 1 - 2 | 99 | 89 | ||
| FQ221516.1 | - | 1 - 2 | 99 | 89 | ||
| FQ221571.1 | - | 1 - 2 | 99 | 89 | ||
| FQ221639.1 | - | 1 - 2 | 99 | 89 | ||
| FQ221768.1 | - | 1 - 2 | 99 | 89 | ||
| FQ222065.1 | - | 1 - 2 | 99 | 89 | ||
| FQ222070.1 | - | 1 - 2 | 99 | 89 | ||
| FQ222149.1 | - | 1 - 2 | 99 | 89 | ||
| FQ222329.1 | - | 1 - 2 | 99 | 89 | ||
| FQ222341.1 | - | 1 - 2 | 89 | |||
| FQ222400.1 | - | 1 - 2 | 99 | 89 | ||
| FQ222420.1 | - | 1 - 2 | 99 | 89 | ||
| FQ222436.1 | - | 1 - 2 | 99 | 89 | ||
| FQ222448.1 | - | 1 - 2 | 99 | 89 | ||
| FQ222605.1 | - | 1 - 2 | 99 | 89 | ||
| FQ222614.1 | - | 1 - 2 | 98 | 89 | ||
| FQ222637.1 | - | 1 - 2 | 99 | 89 | ||
| FQ222674.1 | - | 1 - 2 | 99 | 89 | ||
| FQ222691.1 | - | 1 - 2 | 99 | 89 | ||
| FQ222786.1 | - | 1 - 2 | 99 | 89 | ||
| FQ222845.1 | - | 1 - 2 | 99 | 89 | ||
| FQ222891.1 | - | 1 - 2 | 99 | 89 | ||
| FQ222907.1 | - | 1 - 2 | 99 | 89 | ||
| FQ222910.1 | - | 1 - 2 | 99 | 89 | ||
| FQ222929.1 | - | 1 - 2 | 99 | 89 | ||
| FQ222950.1 | - | 1 - 2 | 99 | 89 | ||
| FQ222957.1 | - | 1 - 2 | 98 | 89 | ||
| FQ223018.1 | - | 1 - 2 | 99 | 89 | ||
| FQ223045.1 | - | 1 - 2 | 99 | 89 | ||
| FQ223170.1 | - | 1 - 2 | 99 | 89 | ||
| FQ223179.1 | - | 1 - 2 | 99 | 89 | ||
| FQ223313.1 | - | 1 - 2 | 99 | 89 | ||
| FQ223425.1 | - | 1 - 2 | 99 | 89 | ||
| FQ223452.1 | - | 1 - 2 | 99 | 89 | ||
| FQ228316.1 | - | 1 - 2 | 99 | 89 | ||
| FQ228334.1 | - | 1 - 2 | 99 | 89 | ||
| FQ228476.1 | - | 1 - 2 | 99 | 89 | ||
| FQ228561.1 | - | 1 - 2 | 99 | 89 | ||
| FQ228573.1 | - | 1 - 2 | 99 | 89 | ||
| FQ228587.1 | - | 1 - 2 | 99 | 89 | ||
| FQ228764.1 | - | 1 - 2 | 99 | 89 | ||
| FQ228920.1 | - | 1 - 2 | 99 | 89 | ||
| FQ228980.1 | - | 1 - 2 | 99 | 89 | ||
| FQ229826.1 | - | 1 - 2 | 99 | 89 | ||
| FQ229982.1 | - | 1 - 2 | 99 | 89 | ||
| J00750.1 | - | 1 - 2 | 98 | 89 | ||
| Gene Symbol: Ttr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | FQ210386.1 | - | 99 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC058442.1,J00750.1,NM_138826.4 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mt1a
Genomic Map