Assay Details
| Gene Symbol: Chchd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108369.1 | NP_001101839.1 | 1 - 2 | 148 | 123 | |
| GenBank mRNA | BC168773.1 | - | 1 - 2 | 144 | 6920789 | 123 |
| FQ217372.1 | - | 1 - 2 | 121 | 123 | ||
| FQ217729.1 | - | 1 - 2 | 128 | 123 | ||
| FQ221927.1 | - | 1 - 2 | 114 | 123 | ||
| FQ224582.1 | - | 1 - 2 | 129 | 123 | ||
| FQ224809.1 | - | 1 - 2 | 129 | 123 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Chchd1
Genomic Map