Assay Details
| Gene Symbol: Med10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106097.2 | NP_001099567.2 | 1 - 2 | 164 | 85 | |
| XM_006227769.2 | XP_006227831.1 | 2 - 3 | 480 | 85 | ||
| GenBank mRNA | CB582092.1 | - | 1 - 2 | 164 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Med10
Genomic Map