Assay Details
Gene Symbol: Rpp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108372.2 | NP_001101842.2 | 5 - 6 | 591 | 66 | |
NM_001141929.1 | NP_001135401.1 | 5 - 6 | 546 | 66 | ||
XM_008770552.1 | XP_008768774.1 | 5 - 6 | 815 | 66 | ||
GenBank mRNA | BU670982.1 | - | 5 - 6 | 592 | 66 | |
CF110818.1 | - | 4 - 5 | 288 | 66 | ||
CK356697.1 | - | 5 - 6 | 546 | 66 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
