Assay Details
| Gene Symbol: Top2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100858.1 | NP_001094328.1 | 1 - 2 | 53 | 69 | |
| GenBank mRNA | AB262979.1 | - | 1 - 2 | 53 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Top2b
Genomic Map