Assay Details
Gene Symbol: Zfp423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053583.2 | NP_446035.2 | 138 | 62 | ||
GenBank mRNA | L03386.1 | - | 138 | 62 | ||
U92564.1 | - | 136 | 62 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the U92564.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
