Assay Details
Gene Symbol: Zfp423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053583.2 | NP_446035.2 | 3945 | 67 | ||
GenBank mRNA | U92564.1 | - | 3943 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
Notch signaling pathway
nervous system development
cell differentiation
positive regulation of BMP signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
nucleic acid binding
protein homodimerization activity
sequence-specific DNA binding
metal ion binding
protein heterodimerization activity
transcription, DNA-templated
Notch signaling pathway
nervous system development
cell differentiation
positive regulation of BMP signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
nucleic acid binding
protein homodimerization activity
sequence-specific DNA binding
metal ion binding
protein heterodimerization activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P055228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698850 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53291 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 29880 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 392107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768555 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 809259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 711223 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212891 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380890 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 195639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 159386 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293951 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 341141 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852617 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625029 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257206 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814695 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 498180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 248656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 540146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694893 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 585395 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184715 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 204932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 884077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 510231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161979 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860163 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 92197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 69898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630754 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 698172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853552 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 938431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 249203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350458 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615359 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304909 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 265809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318610 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 408055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199482 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600558 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 360576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 447373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 730073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 832310 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860593 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 317227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541183 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 283835 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588795 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687282 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 18634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920621 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516145 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 281380 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 197721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 471792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812837 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124487 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733870 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395130 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 550211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229094 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93205 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 73412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565002 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 277962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956335 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 465102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 33019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 861382 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 574248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 712802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 45579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 936723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287822 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 752698 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920262 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 922041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 225498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 86654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28696 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 135277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 160377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 77758 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 138146 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 12041 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 68176 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 21373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 130882 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P463831 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P463831 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P463831 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P463831 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35004 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35161 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53924 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 70050 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 67867 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 20003 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 12761 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37458 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15320 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15141 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 2309 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 89873 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97461 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 64781 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3021 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3002 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 48315 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 39084 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 49277 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 53444 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 91082 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10806342 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806348 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806347 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806350 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806349 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806343 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806345 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806346 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10806341 | - | - |
Rat Genome 230 2.0 Array | 1368542_PM_at | - | - |
Rat Genome 230 2.0 Array | 1396859_PM_at | - | - |
Rat Genome 230 2.0 Array | 1396860_PM_s_at | - | - |
Rat Genome 230 2.0 Array | 1388216_PM_at | - | - |
Rat Genome 230 2.0 Array | 1368542_at | - | - |
Rat Genome 230 2.0 Array | 1396860_s_at | - | - |
Rat Genome 230 2.0 Array | 1396859_at | - | - |
Rat Genome 230 2.0 Array | 1388216_at | - | - |
Affymetrix HT Rat Focus Array | 1396859_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1396860_s_at | - | - |
Affymetrix HT Rat Focus Array | 1368542_at | - | - |
Affymetrix HT Rat Focus Array | 1396859_at | - | - |
Affymetrix HT Rat Focus Array | 1388216_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388216_at | - | - |
Affymetrix HT Rat Focus Array | 1396860_PM_s_at | - | - |
Affymetrix HT Rat Focus Array | 1368542_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1900001725.rn.1 | TC1900000222.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1900001724.rn.1 | TC1900000222.rn.1 | RTA TC |
Rat Expression Set 230 | 1388216_at | - | - |
Rat Expression Set 230 | 1368542_PM_at | - | - |
Rat Expression Set 230 | 1388216_PM_at | - | - |
Rat Expression Set 230 | 1368542_at | - | - |
Rat Expression Set 230 | 1396860_PM_s_at | - | - |
Rat Expression Set 230 | 1396859_at | - | - |
Rat Expression Set 230 | 1396859_PM_at | - | - |
Rat Expression Set 230 | 1396860_s_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10806341 | - | - |
Rat Genome U34 Set | L03386_i_at | - | - |
Rat Genome U34 Set | U92564_g_at | - | - |
Rat Genome U34 Set | U92564_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22241205 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365347 | - | - |
RatRef-12 Expression BeadChip | ILMN_61474 | - | - |
