Assay Details
Gene Symbol: Cyld-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008763560.1 | XP_008761782.1 | 1 | 2884 | 73 |
Gene Symbol: Cyld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017380.1 | NP_001017380.1 | 16 - 17 | 2871 | 73 | |
XM_008772408.1 | XP_008770630.1 | 16 - 17 | 4120 | 73 | ||
GenBank mRNA | AY383658.1 | - | 4 - 5 | 1179 | 73 | |
BC082001.1 | - | 16 - 17 | 2871 | 73 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cytoplasmic translation
ubiquitin-dependent protein catabolic process
regulation of mitotic cell cycle
biological_process
Wnt signaling pathway
protein deubiquitination
negative regulation of NF-kappaB transcription factor activity
negative regulation of NF-kappaB import into nucleus
negative regulation of T cell differentiation
necroptotic process
regulation of microtubule cytoskeleton organization
protein K63-linked deubiquitination
negative regulation of canonical Wnt signaling pathway
ripoptosome assembly involved in necroptotic process
regulation of cilium assembly
positive regulation of extrinsic apoptotic signaling pathway
regulation of intrinsic apoptotic signaling pathway
molecular_function
structural constituent of ribosome
thiol-dependent ubiquitin-specific protease activity
zinc ion binding
protein kinase binding
Lys63-specific deubiquitinase activity
proline-rich region binding
cytoplasmic translation
ubiquitin-dependent protein catabolic process
regulation of mitotic cell cycle
biological_process
Wnt signaling pathway
protein deubiquitination
negative regulation of NF-kappaB transcription factor activity
negative regulation of NF-kappaB import into nucleus
negative regulation of T cell differentiation
necroptotic process
regulation of microtubule cytoskeleton organization
protein K63-linked deubiquitination
negative regulation of canonical Wnt signaling pathway
ripoptosome assembly involved in necroptotic process
regulation of cilium assembly
positive regulation of extrinsic apoptotic signaling pathway
regulation of intrinsic apoptotic signaling pathway
molecular_function
structural constituent of ribosome
thiol-dependent ubiquitin-specific protease activity
zinc ion binding
protein kinase binding
Lys63-specific deubiquitinase activity
proline-rich region binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P116665 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 301047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724405 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338530 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919744 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610454 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162989 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925075 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 746116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588295 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 906340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267713 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86543 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57678 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 117240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107460 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 95401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 72446 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P060456 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P116665 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P060456 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P116665 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P060456 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P116665 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P256261 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 100981 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 96768 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 64232 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10809653 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10809660 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10809659 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10809650 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867421 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867422 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10809646 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10809652 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10809662 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867420 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10809649 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10809654 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867419 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10809645 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10809663 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10809658 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10809651 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10809661 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10809647 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10809664 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867418 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10809648 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867417 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10809656 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10809643 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10867416 | - | - |
Rat Genome 230 2.0 Array | 1392958_PM_at | - | - |
Rat Genome 230 2.0 Array | 1379287_PM_at | - | - |
Rat Genome 230 2.0 Array | 1391091_at | - | - |
Rat Genome 230 2.0 Array | 1391091_PM_at | - | - |
Rat Genome 230 2.0 Array | 1379287_at | - | - |
Rat Genome 230 2.0 Array | 1392958_at | - | - |
Affymetrix HT Rat Focus Array | 1392958_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1392958_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1900008724.rn.1 | TC1900001090.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1900008730.rn.1 | TC1900001090.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1900008725.rn.1 | TC1900001090.rn.1 | RTA TC |
Rat Expression Set 230 | 1392958_at | - | - |
Rat Expression Set 230 | 1392958_PM_at | - | - |
Rat Expression Set 230 | 1379287_at | - | - |
Rat Expression Set 230 | 1391091_at | - | - |
Rat Expression Set 230 | 1379287_PM_at | - | - |
Rat Expression Set 230 | 1391091_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10809643 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10867416 | - | - |
Rat Genome U34 Set | rc_AA891580_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22103009 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1649726 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1374817 | - | - |
