Assay Details
| Gene Symbol: Yae1d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107356.2 | NP_001100826.1 | 1 - 2 | 132 | 99 | |
| GenBank mRNA | FQ216975.1 | - | 1 - 2 | 99 | 99 | |
| Gene Symbol: LOC500028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047954.1 | NP_001041419.1 | 1 | 131 | 99 | |
| GenBank mRNA | AY383683.1 | - | 1 | 131 | 99 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Yae1d1
Genomic Map