Assay Details
Gene Symbol: Cdk13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271295.1 | NP_001258224.1 | 1421 | 56 | ||
NM_001271296.1 | NP_001258225.1 | 1421 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
