Assay Details
Gene Symbol: Ndufb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106360.2 | NP_001099830.1 | 1 - 2 | 92 | 56 | |
GenBank mRNA | BC166889.1 | - | 1 - 2 | 92 | 6891455 | 56 |
BC166905.1 | - | 1 - 2 | 92 | 6887475 | 56 | |
BC166906.1 | - | 1 - 2 | 92 | 6888064 | 56 | |
FQ217125.1 | - | 1 - 2 | 86 | 56 | ||
FQ217646.1 | - | 1 - 2 | 95 | 56 | ||
FQ223847.1 | - | 1 - 2 | 107 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
