Assay Details
| Gene Symbol: Trub1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012173.1 | NP_001012173.1 | 3 - 4 | 425 | 91 | |
| XM_008760538.1 | XP_008758760.1 | 3 - 4 | 456 | 91 | ||
| GenBank mRNA | BC087683.1 | - | 3 - 4 | 425 | 7320660 | 91 |
More Information
Updated Mapping Information
Set Membership: |
|

Trub1
Genomic Map