Assay Details
Gene Symbol: Cela2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012553.2 | NP_036685.1 | 1 - 2 | 59 | 92 | |
GenBank mRNA | FQ226773.1 | - | 1 - 2 | 61 | 92 | |
FQ227652.1 | - | 1 - 2 | 61 | 92 | ||
FQ227675.1 | - | 1 - 2 | 61 | 92 | ||
FQ228000.1 | - | 1 - 2 | 61 | 92 | ||
V01233.1 | - | 1 - 2 | 59 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
