Assay Details
| Gene Symbol: Ptger3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012704.1 | NP_036836.1 | 1 - 2 | 911 | 83 | |
| XM_006233535.2 | XP_006233597.1 | 1 - 2 | 1013 | 83 | ||
| GenBank mRNA | AF302686.1 | - | 1 - 2 | 879 | 83 | |
| D14869.1 | - | 1 - 2 | 879 | 83 | ||
| D16443.1 | - | 1 - 2 | 879 | 83 | ||
| D29969.1 | - | 1 - 2 | 911 | 83 | ||
| X80133.1 | - | 1 - 2 | 928 | 83 | ||
| X83855.1 | - | 1 - 2 | 973 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptger3
Genomic Map