Assay Details
| Gene Symbol: Gucy1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270711.1 | NP_001257640.1 | 7 - 8 | 830 | 65 | |
| NM_012770.2 | NP_036902.2 | 5 - 6 | 713 | 65 | ||
| XM_008770737.1 | XP_008768959.1 | 7 - 8 | 961 | 65 | ||
| XM_008770738.1 | XP_008768960.1 | 7 - 8 | 961 | 65 | ||
| XM_008770739.1 | XP_008768961.1 | 6 - 7 | 1313 | 65 | ||
| XM_008770740.1 | XP_008768962.1 | 6 - 7 | 1090 | 65 | ||
| XM_008770741.1 | XP_008768963.1 | 7 - 8 | 961 | 65 | ||
| GenBank mRNA | AB058888.1 | - | 4 - 5 | 588 | 65 | |
| AB109448.1 | - | 4 - 5 | 567 | 65 | ||
| AB109449.1 | - | 4 - 5 | 567 | 65 | ||
| AB109450.1 | - | 4 - 5 | 567 | 65 | ||
| AB109451.1 | - | 4 - 5 | 567 | 65 | ||
| AY004153.2 | - | 7 - 8 | 830 | 65 | ||
| M57507.1 | - | 5 - 6 | 713 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gucy1b2
Genomic Map