Assay Details
| Gene Symbol: Sirpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013016.2 | NP_037148.2 | 1 - 3 | 317 | 86 | |
| XM_006234951.2 | XP_006235013.1 | 1 - 2 | 86 | |||
| XM_006234952.2 | XP_006235014.1 | 2 - 3 | 86 | |||
| XM_006234953.2 | XP_006235015.1 | 2 - 3 | 86 | |||
| GenBank mRNA | AF055065.1 | - | 2 - 3 | 86 | ||
| BC099773.1 | - | 2 - 3 | 7932298 | 86 | ||
| D38468.1 | - | 1 - 2 | 86 | |||
| D85183.1 | - | 1 - 2 | 86 | |||
| U62328.1 | - | 1 - 3 | 317 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sirpa
Genomic Map