Assay Details
| Gene Symbol: Gjb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019241.1 | NP_062114.1 | 1 | 751 | 66 | |
| XM_008763990.1 | XP_008762212.1 | 2 | 995 | 66 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Gjb5
Genomic Map