Assay Details
| Gene Symbol: Rps21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031111.1 | NP_112373.1 | 1 - 2 | 32 | 55 | |
| XM_006235823.1 | XP_006235885.1 | 1 - 2 | 121 | 55 | ||
| GenBank mRNA | BC058464.1 | - | 1 - 2 | 37 | 6918340 | 55 |
| FQ210654.1 | - | 1 - 2 | 35 | 55 | ||
| FQ210736.1 | - | 1 - 2 | 35 | 55 | ||
| FQ210823.1 | - | 1 - 2 | 33 | 55 | ||
| FQ210868.1 | - | 1 - 2 | 34 | 55 | ||
| FQ212305.1 | - | 1 - 2 | 34 | 55 | ||
| FQ212673.1 | - | 1 - 2 | 27 | 55 | ||
| FQ217300.1 | - | 1 - 2 | 33 | 55 | ||
| FQ221031.1 | - | 1 - 2 | 35 | 55 | ||
| FQ221222.1 | - | 1 - 2 | 35 | 55 | ||
| FQ221352.1 | - | 1 - 2 | 33 | 55 | ||
| FQ221376.1 | - | 1 - 2 | 33 | 55 | ||
| FQ221504.1 | - | 1 - 2 | 33 | 55 | ||
| FQ221719.1 | - | 1 - 2 | 34 | 55 | ||
| FQ221773.1 | - | 1 - 2 | 33 | 55 | ||
| FQ221895.1 | - | 1 - 2 | 33 | 55 | ||
| FQ222195.1 | - | 1 - 2 | 33 | 55 | ||
| FQ222197.1 | - | 1 - 2 | 33 | 55 | ||
| FQ222346.1 | - | 1 - 2 | 34 | 55 | ||
| FQ222395.1 | - | 1 - 2 | 33 | 55 | ||
| FQ222640.1 | - | 1 - 2 | 36 | 55 | ||
| FQ222661.1 | - | 1 - 2 | 33 | 55 | ||
| FQ222692.1 | - | 1 - 2 | 35 | 55 | ||
| FQ222717.1 | - | 1 - 2 | 33 | 55 | ||
| FQ222861.1 | - | 1 - 2 | 33 | 55 | ||
| FQ222985.1 | - | 1 - 2 | 33 | 55 | ||
| FQ223083.1 | - | 1 - 2 | 33 | 55 | ||
| FQ223085.1 | - | 1 - 2 | 33 | 55 | ||
| FQ223126.1 | - | 1 - 2 | 33 | 55 | ||
| FQ223140.1 | - | 1 - 2 | 32 | 55 | ||
| FQ223281.1 | - | 1 - 2 | 33 | 55 | ||
| FQ223324.1 | - | 1 - 2 | 33 | 55 | ||
| FQ223458.1 | - | 1 - 2 | 34 | 55 | ||
| FQ223473.1 | - | 1 - 2 | 63 | 55 | ||
| FQ224512.1 | - | 1 - 2 | 34 | 55 | ||
| FQ224728.1 | - | 1 - 2 | 35 | 55 | ||
| FQ228462.1 | - | 1 - 2 | 33 | 55 | ||
| FQ228565.1 | - | 1 - 2 | 34 | 55 | ||
| FQ228580.1 | - | 1 - 2 | 34 | 55 | ||
| FQ228593.1 | - | 1 - 2 | 33 | 55 | ||
| FQ228604.1 | - | 1 - 2 | 38 | 55 | ||
| FQ229072.1 | - | 1 - 2 | 28 | 55 | ||
| FQ229301.1 | - | 1 - 2 | 37 | 55 | ||
| FQ229813.1 | - | 1 - 2 | 33 | 55 | ||
| X79059.1 | - | 1 - 2 | 32 | 55 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC058464.1,NM_031111.1,X79059.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rps21
Genomic Map